missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EFNB1 (EphrinB1) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of human EFNB1 (NP_004420.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10894H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13446006

  • £56.74 / 5µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Ephrin-B1 (EFNB1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human EFNB1 at 10μg/mL (100μL/well) can bind human EPHB6 with a linear range of 0.16-4μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1947
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
EFNB1, RP3-496G1.1, CFND, CFNS, EFL3, EPLG2, Elk-L, LERK2
EFNB1
Kinases & Inhibitors
His Tag
Lyophilized
24.5kDa
5μg
Truncated/Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin-B1
Assay Standard
Mammalian
Videos