missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EFNA5 (EphrinA5) Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form (Met 1-Asn 203) of mouse EFNA5 long isoform (O08543-1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50597M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13406366

  • £186.34 / 50µg


EU Stock 1
Add to basket

Description

Description

Ephrin A5 (EFNA5), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EPHA5 at 2μg/mL (100μL/well) can bind mouse EFNA4 with a linear range of 1.28-32ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
13640
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS
EFNA5
Kinases & Inhibitors
His Tag
Lyophilized
22.6kDa
50μg
Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin A5
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.