missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EFNA4 (EphrinA4) Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the (Met 1-Gly 176) of mouse EFNA4 (NP_031936.2) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50595M02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13486346

  • £187.38 / 50µg


Estimated Shipment
08-10-2018
Add to basket

Description

Description

Ephrin A4 (EFNA4), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EPHA7 at 2μg/mL (100μL/well) can bind mouse EFNA4-Fc with a linear range of 0.08-10ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
13639
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
AW494418, EFL-2, Ehk1-L, Epl3, LERK-3, mgC129300, mgC129301
EFNA4
Kinases & Inhibitors
IgG1 Fc Tag
Lyophilized
43.9kDa
50μg
Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin A4
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.