Learn More
Thermo Scientific™ Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, hIgG1-Fc Tag
This recombinant protein was expressed from a DNA sequence encoding the mature form of human EphrinA3 (NP_004943.1), residues Met 1 to Ser 213, fused to the Fc region of human IgG1 at the C-terminus
Brand: Thermo Scientific™ 10188H02H250
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Ephrin-A3 (EFNA3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EPHA6 at 1μg/mL (100μL/well) can bind human EFNA3-Fc, the ED50 of human EFNA3 is 299.2ng/mL
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories | |
1944 | |
≥90% by SDS-PAGE | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Ephrins | |
EFNA3, EFL2, EPLG3, Ehk1-L, LERK3 | |
EFNA3 | |
Kinases & Inhibitors | |
IgG1 Fc Tag |
Lyophilized | |
48kDa | |
5 x 50μg | |
Recombinant/Cell Surface Protein/Ligand | |
<1 EU/μg | |
Ephrin-A3 | |
Assay Standard | |
Mammalian |
For Research Use Only. Not for use in diagnostic procedures.