missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EphrinA3 (NP_004943.1), residues Met 1 to Ser 213, fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10188H02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13405776

  • £144.70 / 25µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Ephrin-A3 (EFNA3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EPHA6 at 1μg/mL (100μL/well) can bind human EFNA3-Fc, the ED50 of human EFNA3 is 299.2ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1944
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
EFNA3, EFL2, EPLG3, Ehk1-L, LERK3
EFNA3
Kinases & Inhibitors
IgG1 Fc Tag
Lyophilized
48kDa
5 x 5μg
Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin-A3
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.