missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, DDDDK Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EphrinA3 (NP_004943.1), residues 1 to 213, fused to a DDDDK tag at the C-terminus

Brand:  Thermo Scientific™ 10188HCCH50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15294875

  • £182.18 / 50µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Ephrin-A3 (EFNA3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1944
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
EFNA3, EFL2, EPLG3, Ehk1-L, LERK3
EFNA3
Kinases & Inhibitors
DDDDK
Lyophilized
22kDa
50μg
Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin-A3
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.