Sino Biological™ EDAR Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human EDAR (NP_071731.1) extracellular domain (Met 1-Ile 189) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11577H08H5

EDAR RECOMBINANT HUMAN PROTEIN, HIS TAG

UNSPSC: 12352200

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15248685

Quantity Price
1 £ 54.75 / 5µg
Estimated Shipment date
from Supplier 31-01-2017
Add to basket


Description and Specification

Specification

Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Truncated/Recombinant
Gene Alias DL, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, FLJ94390, HRM1
Validated Application Assay Standard
Expression System Mammalian
Format Lyophilized
Molecular Weight 19kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Common Name Ectodysplasin A receptor
Gene Symbol EDAR
Protein Tag His Tag
Protein Family Cytokines & Receptors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 10913

Ectodysplasin A Receptor (EDAR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.