missing translation for 'onlineSavingsMsg'
Learn More

ECM1 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mouse ECM1 isoform 1 (Q61508-1) (Met 1-Glu 559) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 50331M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15343116

  • £129.55 / 25µg


Estimated Shipment
03-04-2019
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Extracellular Matrix Protein 1 (Ecm1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human mMP-9 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
62.5kDa
5 x 5μg
Glycoproteins
AI663821, p85
ECM1
Other Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Ecm1–extracellular matrix protein 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.