missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ DPP4 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Asn 29-Pro 766) of the mature form of human DPP4 (NP_001926.2) fused to the fused Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10688H01H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15227055

  • £141.58 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Dipeptidyl-Peptidase 4 (DPP4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind recombinant Cynomolgus CXCL12 in a functional ELISA; 2.Measured by its ability to bind recombinant Human SDF1b in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
112kDa
5μg
Proteases
DPP4, ADABP, ADCP2, CD26, DPPIV, TP103
DPP4
Other Proteins
IgG1 Fc Tag
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
DPP4–dipeptidyl-peptidase 4
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.