Sino Biological™ DLL4 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse DLL4 (NP_062327.2) (Met 1-Pro 525) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 50640M08H5

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Includes: One vial of lyophilized protein

Product Code. 15440624

Call for latest availability
+44 (0)1509 555500
Add to basket


Description and Specification

Specification

Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Validated Application Assay Standard
Molecular Weight 55.7kDa
Protein Tag His Tag
Protein Subtype Neurotrophic Factors
Gene Symbol DLL4
Endotoxin Concentration <1 EU/μg
Common Name Delta-like 4 (Drosophila)
Format Lyophilized
Purity ≥90% by SDS-PAGE
Gene Alias Dll4, RP23-46P4.8, Delta4
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Delta-like 4 (Drosophila,) (DLL4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse DLL4-His at 10μg/mL (100μL/well) can bind biotinylated mouse NOTCH1-His; the EC50 of biotinylated mouse NOTCH1-His is 30ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.