missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ DLL1 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human DLL1 (NP_005609.3) extracellular domain (Met 1-Gly 540) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50522M08H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15338896

  • £221.74 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Delta-Like 1 (Drosophila) (DLL1), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human NOTCH1 in a functional ELISA
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, 20% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
55kDa
50μg
Neurotrophic Factors
Delta1
DLL1
Mammalian
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Delta-like 1 (Drosophila)
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.