Learn More
DLL1 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human DLL1 (NP_005609.3) extracellular domain (Met 1-Gly 540) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 50522M08H5
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Delta-Like 1 (Drosophila) (DLL1), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to bind human NOTCH1 in a functional ELISA
- Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, 20% glycerol, pH 7.4
- Reconstitution: No reconstitution needed
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5μg | |
<1 EU/μg | |
Delta-like 1 (Drosophila) | |
His Tag | |
Frozen | |
≥90% by SDS-PAGE |
55kDa | |
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Delta1 | |
DLL1 | |
Mammalian | |
Neurotrophic Factors |
For Research Use Only. Not for use in diagnostic procedures.