Sino Biological™ DLL1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human DLL1 (NP_005609.3) extracellular domain (Met 1-Gly 540) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11635H08H5

UNSPSC: 12352200

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15278755

Quantity Price
1 £ 55.0 / 5µg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Specification

Molecular Weight 57.4kDa
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Common Name Delta-like 1 (Drosophila)
Quantity 5μg
Gene Symbol DLL1
Protein Subtype Neurotrophic Factors
Protein Family Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias DLL1, DELTA1, DL1, Delta
Protein Tag His Tag
Expression System Mammalian
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Delta-Like 1 (Drosophila) (DLL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human NOTCH1 in a functional ELISA
  • Formulation: Lyophilized in 20mM Tris, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.