missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ DLL1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human DLL1 (NP_005609.3) extracellular domain (Met 1-Gly 540) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11635H08H25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15268755

  • £125.97 / 25µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Delta-Like 1 (Drosophila) (DLL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human NOTCH1 in a functional ELISA
  • Formulation: Lyophilized in 20mM Tris, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
57.4kDa
5 x 5μg
Neurotrophic Factors
DLL1, DELTA1, DL1, Delta
DLL1
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Delta-like 1 (Drosophila)
Assay Standard
Mammalian
Videos