missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ DKK3 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 349) of mouse DKK3 (NP_056629.1) precursor fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50247M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15382986

  • £122.84 / 25µg


Estimated Shipment
29-03-2018
Add to basket

Includes: Five vials of protein in suspension buffer

Description

Description

Dickkopf-Related 3 (DKK3), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50781
PAGE
5 x 5μg
C-Terminal
Wnt Proteins & Receptors|Glycoproteins
AW061014, C87148, DKK3
DKK3
Other Proteins
His Tag
Lyophilized
38kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Dickkopf-Related Protein 3
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.