missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ DKK1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human DKK1 precursor (NP_036374.1) (Met 2-His 266) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10170H08H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15294805

  • £110.35 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Dickkopf 1 homolog (Xenopus laevis) (DKK1), recombinant rhesus protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to inhibit WNT3A-induced alkaline phosphatase production by C3H10T1/2 cells; the ED50 for this effect is approximately 0.1-0.4μg/mL in the presence of 100ng/mL of mouse Wnt3a
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
25.8kDa
5μg
Cytokine Receptors
SK, DKK-1
DKK1
Other Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Dickkopf 1 homolog (Xenopus laevis)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.