missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ DDC Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the full length of human DDC (NP_000781.1) (Met 1-Glu 480) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10560H08B5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15276925

  • £133.25 / 5µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Dopa Decarboxylase (DDC), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to convert the substrate 3,4-dihydroxy L-phenylalanine (L-Dopa) to 3,4-dihydroxyphenylethylamine (dopamine); the dopamine product is measured by its absorbance at 340nM after derivatization with trinitrobenzene sulfonic acid, the specific activity is >1500pmole/min./μg
  • Formulation: 50mM Tris, 100mM NaCl, 0.2μg/μL BSA, pH 8, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
55kDa
5μg
Other Proteins
AADC, DDC
DDC
Insect
Frozen
≥85% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Dopa Decarboxylase
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.