missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CXCL9 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human CXCL9 (Q07325) (Thr 23-Thr 125)

Brand:  Thermo Scientific™ 10888HNAE25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15297335

  • £162.81 / 25µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Chemokine (C-X-C Motif) Ligand 9 (CXCL9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4283
PAGE
5 x 5μg
Full Length/Recombinant
<1 EU/μg
Chemokine (C-X-C motif) ligand 9
Assay Standard
E. coli
Lyophilized
11.9kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Other Cytokines
CMK, Humig, MIG, SCYB9, crg-10
CXCL9
Chemokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.