missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CXCL2 Recombinant Mouse Protein, N-His.SUMO Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse CXCL2 (P10889-1) (Ala 28-Asn 100) fused to a N-terminal polyhistide-tagged SUMO tag at the N-terminus

Brand:  Thermo Scientific™ 50070M28E25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15312646

  • £162.81 / 25µg

EU Stock 2
Add to basket

Includes: Five vials of protein in suspension buffer



Chemokine (C-X-C Motif) Ligand 2 (CXCL2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 0.1% TFA, pH 2.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 5μg
GRO/KC/CINC Chemokines|NFκB Pathway Proteins
CINC-2a, GROb, Gro2, MIP-2, MIP-2a, mgsa-b, Mip2, Scyb, Scyb2
Chemokines & Receptors
SUMO Tag, His Tag
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Chemokine (C-X-C motif) ligand 2
Assay Standard
E. coli

For Research Use Only. Not for use in diagnostic procedures.