missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CXCL16 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse CXCL16 (NP_075647.3) extracellular domain (Met 1-Trp 201) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50142M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15302806

  • £142.07 / 25µg


Estimated Shipment
11-05-2018
Add to basket

Includes: Five vials of protein in suspension buffer

Description

Description

Chemokine (C-X-C Motif) Ligand 16 (CXCL16), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
66102
PAGE
5 x 5μg
C-Terminal
Other Cytokines
DN-491F10.6, 0910001K24Rik, AV290116, BB024863, SR-PSOX, Zmynd15, CXCL16
CXCL16
Chemokines & Receptors
His Tag
Lyophilized
20.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Chemokine (C-X-C motif) ligand 16
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.