Learn More
Thermo Scientific™ Sino Biological™ CXCL12 Recombinant Human Protein
This recombinant protein was expressed from a DNA sequence encoding the human CXCL12 isoform beta (P48061-1) (Lys 22- Met 93)
Brand: Thermo Scientific™ 10118HNAE50
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Chemokine (C-X-C Motif) Ligand 12 (CXCL12), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: The activity of this protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories | |
6387 | |
PAGE | |
50μg | |
Truncated/Recombinant | |
SCYB12, SDF-1a, SDF-1b, SDF1, SDF1A, SDF1B, TLSF-a, TLSF-b, TPAR1 | |
CXCL12 | |
Chemokines & Receptors |
Lyophilized | |
8.6kDa | |
≥90% by SDS-PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
<1 EU/μg | |
Chemokine (C-X-C motif) ligand 12 | |
Assay Standard | |
E. coli |
For Research Use Only. Not for use in diagnostic procedures.