missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CXCL12 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human CXCL12 isoform beta (P48061-1) (Lys 22- Met 93)

Brand:  Thermo Scientific™ 10118HNAE5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15294675

  • £83.08 / 5µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Chemokine (C-X-C Motif) Ligand 12 (CXCL12), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6387
PAGE
5μg
Truncated/Recombinant
<1 EU/μg
Chemokine (C-X-C motif) ligand 12
Assay Standard
E. coli
Lyophilized
8.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
SDFs (Stromal Cell-Derived Factors)
SCYB12, SDF-1a, SDF-1b, SDF1, SDF1A, SDF1B, TLSF-a, TLSF-b, TPAR1
CXCL12
Chemokines & Receptors
Videos