missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CTSS Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the pro form of human CTSS (NP_004070.3) (Met 1-Ile 331) fused to a carboxy-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10487H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15256815

  • £415.36 / 25µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Cathepsin S (CTSS), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate, (7-methoxycoumarin-4-yl) acetyl-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys (2, 4-dinitrophenyl)-NH2 (R∓D Systems Cat. No. ES002); cleavage of ES002 can be measured using excitation and emission wavelengths at 320nM and 405nM, respectively, the specific activity is >300pmoles/min./μg
  • Formulation: 12.5mM MES, 0.075M NaCl, 10% glycerol, pH 6.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
37kDa
5 x 5μg
Proteases
MGC3886, CTSS
CTSS
Mammalian
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Cathepsin S
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.