Sino Biological™ CTSL1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the pro form of human CTSL1 (NP_001903.1) (Met 1-Val 333) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10486H08H50

CTSL1 RECOMBINANT HUMAN PROTEIN, HIS TAG

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15246815

Quantity Price
1 £ 726.0 / 50µg
Estimated Shipment date
from Supplier 01-02-2017
Add to basket


Description and Specification

Specification

Purity ≥85% by SDS-PAGE
Quantity 50μg
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Molecular Weight 37.3kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Proteases|Glycoproteins
Gene Alias CTSL1, CATL, CTSL, FLJ31037, MEP
Common Name Cathepsin L1
Gene Symbol CTSL1
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Cathepsin L1 (CTSL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD74 at 5μg/mL (100μL/well) can bind biotinylated human CTSL1 with a linear range of 3.2-400ng/mL
  • Formulation: Lyophilized in 50mM NaAc, 0.1M NaCl, pH 5.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.