missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CTSL1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the pro form of human CTSL1 (NP_001903.1) (Met 1-Val 333) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10486H08H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15236815

  • £138.46 / 5µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Cathepsin L1 (CTSL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD74 at 5μg/mL (100μL/well) can bind biotinylated human CTSL1 with a linear range of 3.2-400ng/mL
  • Formulation: Lyophilized in 50mM NaAc, 0.1M NaCl, pH 5.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
37.3kDa
5μg
Proteases|Glycoproteins
CTSL1, CATL, CTSL, FLJ31037, MEP
CTSL1
Other Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Cathepsin L1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.