missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CTSB Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the pre pro form of human CTSB (NP_001899.1) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50084M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15382676

  • £139.50 / 5µg


Estimated Shipment
08-06-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

Cathepsin B (CTSB), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate Z-LR-AMC (R&D Systems Cat. No. ES008); the specific activity is >2000pmole/min./μg
  • Formulation: Lyophilized in 20mM Tris, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
36.6kDa
5μg
Proteases
CB
CTSB
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Cathepsin B
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.