missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CTSA Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human cathepsin A isoform b (Met 1-Tyr 480) (NP_001121167.1) fused to a N-terminal signal peptide and a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10482H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15296795

  • £142.62 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Cathepsin A (CTSA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5476
PAGE
5μg
C-Terminal
Proteases|Peptidase
CTSA, RP3-337O18.1, GLB2, GSL, NGBE, PPCA, PPGB
CTSA
Proteases, Peptidases & Inhibitors
His Tag
Lyophilized
53kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Cathepsin A
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.