Learn More
CTLA4 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CTLA4 (NP_033973.2) (Met 1-Phe 162) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 50503M08H250
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: Five vials of protein in suspension buffer
Description
Cytotoxic T-Lymphocyte-Associated 4 (CTLA4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized recombinant mouse CTLA4-His (Mfr. No. 50503-M08H) at 10μg/mL (100μL/well) can mouse CD80-Fc (Mfr. No. 50446-M02H) with a linear range of 0.78-62.5ng/mL; 2. Immobilized recombinant mouse CTLA4-His (Mfr. No. 50503-M08H) at 10μg/mL (100μL/well) can human B7-1-Fc (Mfr. No. 10698-H03H) with a linear range of 0.78-125ng/mL
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
15.3kDa | |
5 x 50μg | |
C-Terminal | |
<1 EU/μg | |
Cytotoxic T-lymphocyte-associated protein 4 | |
His Tag | |
CD Proteins | |
≥90% by SDS-PAGE |
12477 | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
Truncated/Recombinant | |
CH29-120A16.2, Cd152, Ctla-4, Ly-56, Ctla4 | |
CTLA4 | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.