missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CTLA4 Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CTLA4 (NP_033973.2) (Met 1-Phe 162) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50503M02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15388866

  • £57.00 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Cytotoxic T-Lymphocyte-Associated Protein 4 (CTLA4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12477
PAGE
5μg
C-Terminal
<1 EU/μg
Cytotoxic T-lymphocyte-associated protein 4
Assay Standard
Mammalian
Lyophilized
41kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
CH29-120A16.2, Cd152, Ctla-4, Ly-56, Ctla4
CTLA4
CD Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.