Sino Biological™ CSTA Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CSTA (NP_003801.1) (Thr 95-Gly 281) fused to a polyhistidinetag at the N-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10479H07E5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15236785

Quantity Price
1 £ 55.0 / 5µg
Estimated Shipment date
from Supplier 15-03-2017
Add to basket


Description and Specification

Specification

Molecular Weight 22.6kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Endotoxin Concentration <1 EU/μg
Gene ID (Entrez) 1475
Structural Form Full Length/Recombinant
Gene Alias STF1, STFA
Purity ≥90% by SDS-PAGE
Expression System E. coli
Protein Tag His Tag
Protein Family Proteases, Peptidases & Inhibitors
Validated Application Assay Standard
Format Lyophilized
Termination N-Terminal
Purification Method PAGE
Common Name Cystatin A
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Symbol CSTA
Quantity 5μg

Cystatin-A (CSTA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 1mM EDTA, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.