missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CSTA Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CSTA (NP_003801.1) (Thr 95-Gly 281) fused to a polyhistidinetag at the N-terminus

Brand:  Thermo Scientific™ 10479H07E25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15216785

  • £127.01 / 25µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Cystatin-A (CSTA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 1mM EDTA, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1475
PAGE
5 x 5μg
N-Terminal
<1 EU/μg
Cystatin A
Assay Standard
E. coli
Lyophilized
22.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
STF1, STFA
CSTA
Proteases, Peptidases & Inhibitors
His Tag
Videos