Sino Biological™ CSH1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CSH1 (NP_001308.1) (Met 1-Phe 217) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11596H08H25

UNSPSC: 12352200

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15208705

Quantity Price
1 £ 217.0 / 25µg
EU Stock 2
Add to basket


Description and Specification

Specification

Protein Tag His Tag
Termination C-Terminal
Molecular Weight 23.7kDa
Quantity 5 x 5μg
Format Lyophilized
Protein Family Hormones & Receptors
Purification Method PAGE
Gene ID (Entrez) 1442
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Expression System Mammalian
Purity ≥85% by SDS-PAGE
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias CSH1
Common Name Chorionic somatomammotropin hormone
Gene Symbol CSH1
Validated Application Assay Standard
Structural Form Full Length/Recombinant
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Chorionic Somatomammotropin Hormone (CSH1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.