missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CSH1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CSH1 (NP_001308.1) (Met 1-Phe 217) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11596H08H50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238705

  • £354.99 / 50µg


Estimated Shipment
08-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Chorionic Somatomammotropin Hormone (CSH1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1442
PAGE
50μg
C-Terminal
<1 EU/μg
Chorionic somatomammotropin hormone
Assay Standard
Mammalian
Lyophilized
23.7kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
CSH1
CSH1
Hormones & Receptors
His Tag
Videos