missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CSF3R (CD114) Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 621) of human CSF3R precursor (NP_000751.1) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10218H02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13445816

  • £144.70 / 25µg


Estimated Shipment
06-07-2018
Add to basket

Description

Description

Colony stimulating factor 3 receptor (CSF3R/CD114), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard or in other research applications.
  • Activity: Measured by its ability to inhibit the GCSF-induced proliferation of NFS-60 mouse myeloid cells, the ED50 for this effect is typically 1-4ng/mL in the presence of 0.125ng/mL of recombinant human GCSF
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1441
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Cytokine Receptors
CD114, GCSFR, CSF3R
CSF3R
Cytokines & Receptors/CD Proteins
IgG1 Fc Tag
Lyophilized
93.3kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Colony stimulating factor 3 receptor (granulocyte)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.