missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CSF2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human CSF2 (NP_000749.2) (Ala 18-Glu 144) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10015H07H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13472919

  • £255.05 / 50µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Colony stimulating factor 2 (granulocyte-macrophage) (CSF2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF-1 human erythroleukemic cells; the ED50 for this effect is typically 0.1-0.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16.9kDa
50μg
Recombinant/Inactive
<1 EU/μg
Colony stimulating factor 2 (granulocyte-macrophage)
Assay Standard
Mammalian
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Colony Stimulating Factors
CSF2, GMCSF, mgC131935, mgC138897
CSF2
Cytokines & Receptors
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.