missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CSF1R (MCSFR) Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 511) of mouse CSF1R (NP_001032948.2) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50059M08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13425636

  • £920.25 / 250µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to bind recombinant mouse IL34 in a functional ELISA; immobilized mouse CSF1R-His at 10μg/mL (100μL/well) can bind biotinylated human CSF1-His; the ED50 of biotinylated human CSF1-His is 0.50μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12978
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Cytokine Receptors
ICsf1r, AI323359, CD115, CSF-1R, Csfmr, Fim-2, Fms, M-CSFR
CSF1R
Growth Factors & Receptors
His Tag
Lyophilized
56.8kDa
5 x 50μg
Recombinant/Cell Surface Protein
<1 EU/μg
Colony stimulating factor 1 receptor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.