missing translation for 'onlineSavingsMsg'
Learn More

CSF1R (MCSFR) Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu512) of human CSF1R precursor (NP_005202.2) fused to the a C-terminal polyhistidine tag

Brand:  Invitrogen 10161H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13435746

  • £568.00 / 250µg

Estimated Shipment date
from Supplier 27-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R at 10μg/mL can bind biotinylated human CSF1, the ED50 of biotinylated human CSF1 is 8ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1436
>98% by SDS-PAGE
Store at -20 to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Cytokine Receptors
M-CSF R, CSF1R, FMS, CSFR, FIM2, C-FMS, CD115
CSF1R
Growth Factors & Receptors
His Tag
Lyophilized
56kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Colony stimulating factor 1 receptor
Assay Standard
Mammalian
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.