Sino Biological™ CSF1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Met 1-Asn 190) of human CSF1 (P09603-1) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11792H08H5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00001kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 13403069

Quantity Price
1 £ 85.75 / 5µg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Specification

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Lyophilized
Gene Symbol CSF1
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5μg
Common Name Colony stimulating factor 1 (macrophage)
Expression System Mammalian
Molecular Weight 19.8kDa
Structural Form Truncated/Recombinant/Cell Surface Protein
Endotoxin Concentration <1 EU/μg
Protein Subtype Colony Stimulating Factors
Protein Family Cytokines & Receptors
Validated Application Assay Standard
Protein Tag His Tag
Purity >98% by SDS-PAGE
Gene Alias RP11-195M16.2, MCSF, mgC31930

Macrophage Colony Stimulating Factor (CSF1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R (100μL/well of a 10μg/mL solution) can bind biotinylated human CSF1; the EC50 of biotinylated human CSF1 is 8ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.