Learn More
Thermo Scientific™ Sino Biological™ CRP Recombinant Human Protein, His Tag
This recombinant protein was expressed from a DNA sequence encoding human CRP (NP_000558.2) (Met 1-Pro 224) fused to a polyhistidine tag at the C-terminus
Brand: Thermo Scientific™ 11250H08H25
Additional Details : Weight : 0.10000kg
Description
C-Reactive Protein Pentraxin-Related (CRP) recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to bind human CD32a (Mfr. No.10374-H08B) in functional ELISA
- Formulation: Lyophilized in 20mM Tris, 0.2M NaCl, 5mM CaCl2, pH 8.0
- Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories | |
1401 | |
PAGE | |
5 x 5μg | |
C-Terminal | |
RP11-419N10.4, mgC149895, mgC88244, PTX1 | |
CRP | |
Other Proteins | |
His Tag |
Lyophilized | |
24.7kDa | |
≥90% by SDS-PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Full Length/Recombinant | |
C-reactive protein, pentraxin-related | |
Assay Standard | |
Mammalian |
For Research Use Only. Not for use in diagnostic procedures.