Sino Biological™ CPA2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the pre pro form of human CPA2 (NP_001859.1) (Met 1-Tyr 417) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10499H08H50

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15266825

Quantity Price
1 £ 678.0 / 50µg
Estimated Shipment date
from Supplier 15-05-2017
Add to basket


Description and Specification

Specification

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 50μg
Molecular Weight 46kDa
Purity ≥85% by SDS-PAGE
Gene Alias CPA2
Gene Symbol CPA2
Endotoxin Concentration <1 EU/μg
Expression System Mammalian
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Protein Family Other Proteins
Protein Tag His Tag
Common Name Carboxypeptidase A2 ( CPA2 ) Protein
Protein Subtype Proteases

Carboxypeptidase A2 (pancreatic) (CPA2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave a colorimetric peptide substrate, N-acetyl-Phe-Thiaphe-OH (N-Ac-PSP, Peptide International Cat. No. STP-3621-PI), in the presence of 5,5'Dithio-bis (2-nitrobenzoic acid) (DTNB), as measured using the wavelength at 405nM and the extinction coefficient of 13,260 M-1 cm-1, the specific activity is >4nM/min./μg
  • Formulation: Lyophilized in 25mM Tris, 0.15M NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.