missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CPA1 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse CPA1 precusor (NP_079626.2) (Met 1-Tyr 419) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50448M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15313246

  • £722.46 / 50µg

Estimated Shipment
Add to basket

Includes: One vial of lyophilized protein



Carboxypeptidase A1, pancreatic (CPA1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the colorimetric peptide substrate Ac-Phe-Thiaphe-OH in the presence of 5,5'Dithiobis (2-nitrobenzoic acid) (DTNB); the specific activity is >25nM/min./μg
  • Formulation: Lyophilized in 20mM Tris, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
CPA1, 0910001L12Rik, Cpa
Other Proteins
His Tag
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Carboxypeptidase A1, pancreatic
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.