missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CNTNAP2 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CNTNAP2 (NP_001004357.2) (Met 1-Ser 1262) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50649M08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15329096

  • £139.50 / 25µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer

Description

Description

Contactin Associated Protein-like 2 (CNTNAP2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of C6 Rat brain glial cells; mouse CNTNAP2 immobilized at 0.8μg/mL, 100μL/well will mediate >30% C6 cell adhesion
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
66797
PAGE
5 x 5μg
C-Terminal
Other Proteins
CNTNAP2, 5430425M22Rik, Caspr2, Mkiaa0868
CNTNAP2
Other Proteins
His Tag
Lyophilized
139kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
<1 EU/μg
Contactin associated protein-like 2
Assay Standard
Mammalian
Videos