missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ CLMP (ASAM) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of the human ASAM precursor (NP_079045.1) (Met 1-Met 233) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 10794H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 15267175

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

CXADR-like membrane protein (ASAM), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line; when 4 x 104 cells/well are added to human ASAM coated plates (30μg/mL, 100μL/well), approximately 25-50 % will adhere after one hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
25.8kDa
5 x 5μg
Other Proteins
ASAM, ACAM, CLMP, FLJ22415
CLMP
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CXADR-like membrane protein
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.