missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CLEC12A (MICL) Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CLEC12A (EAW96132.1) extracellular domain (His 75-Ala 275) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11896H07H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15205595

  • £637.10 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

C-type Lectin domain family 12, member A (CLEC12A), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 7.4
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
160364
PAGE
50μg
N-Terminal
<1 EU/μg
C-type Lectin domain family 12, member A
Assay Standard
Mammalian
Lyophilized
26kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
CLL-1, CLL1, DCAL-2, mgC70602, MICL
CLEC12A
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.