missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CHI3L1 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CHI3L1 (Q61362) (Met 1-Ala 381) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50929M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15379316

  • £201.96 / 50µg


Estimated Shipment
25-06-2018
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Chitinase 3-like 1 (CHI3L1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12654
PAGE
50μg
C-Terminal
<1 EU/μg
Chitinase 3-like 1
Assay Standard
Mammalian
Lyophilized
42.3kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
AW208766, Brp39, Gp39
CHI3L1
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.