missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CHI3L1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CHI3L1 (NP_001267.2) (Met 1-Thr 383) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11227H08H5

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15238425

  • £55.51 / 5µg


Estimated Shipment
05-06-2018
Add to basket

Description

Description

Chitinase 3-Like 1 (Cartilage Glycoprotein-39) (CHI3L1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of Fadu human squamous cell carcinoma cells (ATCC: HTB43); when cells are added to recombinant human CHI3L1 coated plates (1.5μg/mL, 100μL/well), approximately 65-75% will adhere specifically after 30 minutes at 37°C
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
42kDa
5μg
Other Proteins
ASRT7, DKFZp686N19119, FLJ38139, GP39, HC-gp39, HCGP-3P, YKL40, YYL-40
CHI3L1
Other Proteins
His Tag
Frozen
≥95% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Chitinase 3-like 1 (cartilage glycoprotein-39)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.