Learn More
CFD (Adipsin) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of human CFD (P00746) (Ile 26-Ala 253) fused to a C-terminal polyhistidine tag and a signal peptide at the N-terminus
Brand: Invitrogen 10160H08H5
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Complement Factor D (CFD), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: The activity of this protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
25.8kDa | |
5μg | |
C-Terminal | |
<1 EU/μg | |
Complement factor D (adipsin) | |
His Tag | |
Other Proteins | |
Proteases, Complement, Glycoproteins |
1675 | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
Full Length/Recombinant | |
ADIPSIN, ADN, DF, PFD | |
CFD | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.