missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM6 (CD66C) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human CEACAM6 (NP_002474.3) (Met 1-Gly 320) fused to a polyhistidine tag at the C-terminus
£57.25
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 32.6kDa |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Endotoxin Concentration | <1 EU/μg |
Gene Alias | CEACAM6, CD66c, CEAL, NCA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15257245
|
Invitrogen
10823H08H5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Carcinoembryonic antigen-related cell adhesion molecule 6 (nonspecific cross reacting antigen) (CEACAM6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: 1. Measured by its binding ability in a functional ELISA; 2. Immobilized human CEACAM6-His (Mfr. No. 10823-H08H) at 10μg/mL (100μL/well) can bind biotinylated human CEACAM8-His, the EC50 of biotinylated human CEACAM8-His is 0.17μg/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
CEACAM6, CD66c, CEAL, NCA | |
CEACAM6 | |
Mammalian | |
Lyophilized |
32.6kDa | |
<1 EU/μg | |
Carcinoembryonic antigen-related cell adhesion molecule 6 | |
His Tag | |
CD Proteins | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.