missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CEACAM6 (CD66C) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CEACAM6 (NP_002474.3) (Met 1-Gly 320) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10823H08H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15247245

  • £801.57 / 250µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Carcinoembryonic antigen-related cell adhesion molecule 6 (nonspecific cross reacting antigen) (CEACAM6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; 2. Immobilized human CEACAM6-His (Mfr. No. 10823-H08H) at 10μg/mL (100μL/well) can bind biotinylated human CEACAM8-His, the EC50 of biotinylated human CEACAM8-His is 0.17μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
32.6kDa
5 x 50μg
<1 EU/μg
Carcinoembryonic antigen-related cell adhesion molecule 6
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CEACAM6, CD66c, CEAL, NCA
CEACAM6
CD Proteins
His Tag
Videos