Learn More
Thermo Scientific™ Sino Biological™ CEACAM1 Recombinant Mouse Protein, His Tag
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CEACAM1 (NP_001034274.1) (Met 1-Gly 428) fused to a polyhistidine tag at the C-terminus
Brand: Thermo Scientific™ 50571M08H50
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: One vial of protein in suspension buffer
Description
Carcinoembryonic Antigen-Related Cell Adhesion Molecule 1 (CEACAM1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: The activity of this protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories | |
26365 | |
PAGE | |
50μg | |
C-Terminal | |
<1 EU/μg | |
Carcinoembryonic antigen-related cell adhesion molecule 1 | |
Assay Standard | |
Mammalian |
Lyophilized | |
45kDa | |
≥90% by SDS-PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
Truncated/Recombinant | |
Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1 | |
CEACAM1 | |
CD Proteins | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.